Anti-NOL3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305985EA 600 EUR
ORIGTA305985
Anti-NOL3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NOL3 (nucleolar protein 3 (apoptosis repressor with CARD domain))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR