Anti-CASP9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305983EA 588 EUR
ORIGTA305983
Anti-CASP9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000 Immunochemistry: 2 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: CASP9 (caspase 9, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR