Anti-MADD Rabbit Polyclonal Antibody

Lieferant: OriGene
TA305958
ORIGTA305958EA 600 EUR
ORIGTA305958
Anti-MADD Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 mg/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: MADD (MAP-kinase activating death domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR