Anti-DFFA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305957EA 600 EUR
ORIGTA305957
Anti-DFFA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: DFFA (DNA fragmentation factor, 45kDa, alpha polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR