Anti-ADAM17 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305951EA 588 EUR
ORIGTA305951
Anti-ADAM17 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5 μg/ml;; Immunofluorescence: 10 μg/ml

Type: Primary
Antigen: ADAM17 (ADAM metallopeptidase domain 17)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR