Anti-CRADD Rabbit Polyclonal Antibody

Lieferant: OriGene
TA305943
ORIGTA305943EA 600 EUR
ORIGTA305943
Anti-CRADD Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: CRADD (CASP2 and RIPK1 domain containing adaptor with death domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR