Anti-TCHHL1 Goat Antibody

Lieferant: OriGene
ORIGTA305867EA 547 EUR
ORIGTA305867
Anti-TCHHL1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: TCHHL1 (trichohyalin-like 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR