Anti-UNC5CL Goat Antibody

Lieferant: OriGene
ORIGTA305863EA 547 EUR
ORIGTA305863
Anti-UNC5CL Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: UNC5CL (unc-5 homolog C (C. elegans)-like)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR