Anti-EPM2A Goat Antibody

Lieferant: OriGene
ORIGTA305859EA 547 EUR
ORIGTA305859
Anti-EPM2A Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR