Anti-ERN1 Goat Antibody

Lieferant: OriGene
ORIGTA305857EA 536 EUR
ORIGTA305857
Anti-ERN1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: ERN1 (endoplasmic reticulum to nucleus signaling 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR