Anti-SPHK1 Goat Antibody

Lieferant: OriGene
ORIGTA305846EA 539 EUR
ORIGTA305846
Anti-SPHK1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-3?g/ml

Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR