Anti-DLG1 Goat Antibody

Lieferant: OriGene
ORIGTA305843EA 536 EUR
ORIGTA305843
Anti-DLG1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: DLG1 (discs, large homolog 1 (Drosophila))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR