Anti-ADH5 Goat Antibody

Lieferant: OriGene
ORIGTA305835EA 547 EUR
ORIGTA305835
Anti-ADH5 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ADH5 (alcohol dehydrogenase 5 (class III), chi polypeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR