Anti-ZDHHC13 Goat Antibody

Lieferant: OriGene
ORIGTA305825EA 536 EUR
ORIGTA305825
Anti-ZDHHC13 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ZDHHC13 (zinc finger, DHHC-type containing 13)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR