Anti-KDM2B Goat Antibody

Lieferant: OriGene
ORIGTA305816EA 547 EUR
ORIGTA305816
Anti-KDM2B Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: KDM2B (lysine (K)-specific demethylase 2B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR