Anti-CARD6 Goat Antibody

Lieferant: OriGene
ORIGTA305815EA 550 EUR
ORIGTA305815
Anti-CARD6 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: CARD6 (caspase recruitment domain family, member 6)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR