Anti-FEM1A Goat Antibody

Lieferant: OriGene
ORIGTA305810EA 550 EUR
ORIGTA305810
Anti-FEM1A Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: FEM1A (fem-1 homolog a (C. elegans))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR