Anti-ALS2 Goat Antibody

Lieferant: OriGene
ORIGTA305806EA 536 EUR
ORIGTA305806
Anti-ALS2 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: ALS2 (amyotrophic lateral sclerosis 2 (juvenile))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR