Anti-NDEL1 Goat Antibody

Lieferant: OriGene
ORIGTA305793EA 536 EUR
ORIGTA305793
Anti-NDEL1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: NDEL1 (nudE neurodevelopment protein 1-like 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR