Anti-CAV3 Goat Antibody

Lieferant: OriGene
ORIGTA305791EA 536 EUR
ORIGTA305791
Anti-CAV3 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: CAV3 (caveolin 3)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR