Anti-HMGA1 Goat Antibody

Lieferant: OriGene
ORIGTA305780EA 547 EUR
ORIGTA305780
Anti-HMGA1 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: HMGA1 (high mobility group AT-hook 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR