Anti-TNFSF14 Goat Antibody

Lieferant: OriGene
ORIGTA305766EA 536 EUR
ORIGTA305766
Anti-TNFSF14 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: TNFSF14 (tumor necrosis factor (ligand) superfamily, member 14)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR