Anti-MTHFS Goat Antibody

Lieferant: OriGene
ORIGTA305750EA 539 EUR
ORIGTA305750
Anti-MTHFS Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: MTHFS (5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR