Anti-POLL Goat Antibody

Lieferant: OriGene
ORIGTA305748EA 547 EUR
ORIGTA305748
Anti-POLL Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: POLL (polymerase (DNA directed), lambda)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR