Anti-HMG20A Goat Antibody

Lieferant: OriGene
ORIGTA305740EA 547 EUR
ORIGTA305740
Anti-HMG20A Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1-2mg/ml

Type: Primary
Antigen: HMG20A (high mobility group 20A)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR