Anti-MEPCE Goat Antibody

Lieferant: OriGene
ORIGTA305736EA 550 EUR
ORIGTA305736
Anti-MEPCE Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: MEPCE (methylphosphate capping enzyme)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR