Anti-GOPC Goat antibody

Lieferant: OriGene
ORIGTA305733EA 536 EUR
ORIGTA305733
Anti-GOPC Goat antibody
Antikörper
Recommended Dilutions: Western Blot: 005-02?g/ml

Type: Primary
Antigen: GOPC (golgi-associated PDZ and coiled-coil motif containing)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR