Anti-ITM2B Goat Antibody

Lieferant: OriGene
ORIGTA305731EA 547 EUR
ORIGTA305731
Anti-ITM2B Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ITM2B (integral membrane protein 2B)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR