Anti-PRUNE2 Goat Antibody

Lieferant: OriGene
ORIGTA305724EA 547 EUR
ORIGTA305724
Anti-PRUNE2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: PRUNE2 (prune homolog 2 (Drosophila))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR