Anti-F11R Goat antibody

Lieferant: OriGene
ORIGTA305723EA 547 EUR
ORIGTA305723
Anti-F11R Goat antibody
Antikörper
Recommended Dilutions: Western Blot: 03-2?g/ml

Type: Primary
Antigen: F11R (F11 receptor)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR