Anti-SEPT6 Goat Antibody

Lieferant: OriGene
ORIGTA305718EA 536 EUR
ORIGTA305718
Anti-SEPT6 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.03-1?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:4000

Type: Primary
Antigen: SEPT6 (septin 6)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Bovine, Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR