Anti-PIAS2 Goat Antibody

Lieferant: OriGene
ORIGTA305708EA 547 EUR
ORIGTA305708
Anti-PIAS2 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 3-5mg/ml

Type: Primary
Antigen: PIAS2 (protein inhibitor of activated STAT, 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR