Anti-RASSF3 Goat Antibody

Lieferant: OriGene
ORIGTA305705EA 550 EUR
ORIGTA305705
Anti-RASSF3 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: RASSF3 (Ras association (RalGDS/AF-6) domain family member 3)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR