Anti-MBNL1 Goat Antibody

Lieferant: OriGene
ORIGTA305699EA 547 EUR
ORIGTA305699
Anti-MBNL1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: MBNL1 (muscleblind-like splicing regulator 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR