Anti-FBXO32 Goat antibody

Lieferant: OriGene
ORIGTA305697EA 536 EUR
ORIGTA305697
Anti-FBXO32 Goat antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: FBXO32 (F-box protein 32)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR