Anti-RAPSN Goat Antibody

Lieferant: OriGene
ORIGTA305694EA 536 EUR
ORIGTA305694
Anti-RAPSN Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: RAPSN (receptor-associated protein of the synapse)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR