Anti-ZEB1 Goat Antibody

Lieferant: OriGene
ORIGTA305688EA 536 EUR
ORIGTA305688
Anti-ZEB1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 005-02?g/ml

Type: Primary
Antigen: ZEB1 (zinc finger E-box binding homeobox 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR