Anti-PNPLA2 Goat Antibody

Lieferant: OriGene
ORIGTA305683EA 547 EUR
ORIGTA305683
Anti-PNPLA2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-2?g/ml

Type: Primary
Antigen: PNPLA2 (patatin-like phospholipase domain containing 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR