Anti-SAR1A Goat Antibody

Lieferant: OriGene
ORIGTA305682EA 547 EUR
ORIGTA305682
Anti-SAR1A Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: SAR1A (secretion associated, Ras related GTPase 1A)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR