Anti-TRPC4 Goat Antibody

Lieferant: OriGene
ORIGTA305678EA 547 EUR
ORIGTA305678
Anti-TRPC4 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: TRPC4 (transient receptor potential cation channel, subfamily C, member 4)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR