Anti-NUAK1 Goat Antibody

Lieferant: OriGene
ORIGTA305675EA 550 EUR
ORIGTA305675
Anti-NUAK1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: NUAK1 (NUAK family, SNF1-like kinase, 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR