Anti-KPNA6 Goat Antibody

Lieferant: OriGene
ORIGTA305673EA 536 EUR
ORIGTA305673
Anti-KPNA6 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: KPNA6 (karyopherin alpha 6 (importin alpha 7))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR