Anti-ATF5 Goat antibody

Lieferant: OriGene
ORIGTA305671EA 536 EUR
ORIGTA305671
Anti-ATF5 Goat antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ATF5 (activating transcription factor 5)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR