Anti-RNF139 Goat Antibody

Lieferant: OriGene
ORIGTA305669EA 536 EUR
ORIGTA305669
Anti-RNF139 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05?g/ml

Type: Primary
Antigen: RNF139 (ring finger protein 139)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR