Anti-ANXA10 Goat Antibody

Lieferant: OriGene
ORIGTA305668EA 539 EUR
ORIGTA305668
Anti-ANXA10 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 3-5mg/ml

Type: Primary
Antigen: ANXA10 (annexin A10)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR