Anti-HAX1 Goat antibody

Lieferant: OriGene
ORIGTA305662EA 547 EUR
ORIGTA305662
Anti-HAX1 Goat antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: HAX1 (HCLS1 associated protein X-1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR