Anti-LIPG Goat Antibody

Lieferant: OriGene
ORIGTA305661EA 547 EUR
ORIGTA305661
Anti-LIPG Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: LIPG (lipase, endothelial)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR