Anti-NR1I2 Goat Antibody

Lieferant: OriGene
ORIGTA305651EA 547 EUR
ORIGTA305651
Anti-NR1I2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: NR1I2 (nuclear receptor subfamily 1, group I, member 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR