Anti-DPM1 Goat Antibody

Lieferant: OriGene
ORIGTA305650EA 536 EUR
ORIGTA305650
Anti-DPM1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: DPM1 (dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR