Anti-SFRP4 Goat Antibody

Lieferant: OriGene
ORIGTA305645EA 536 EUR
ORIGTA305645
Anti-SFRP4 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml; Immunochemistry: 2-4?g/ml; ELISA: 1:32000

Type: Primary
Antigen: SFRP4 (secreted frizzled-related protein 4)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR